Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289514.1 | NP_001276443.1 | 10 | 1524 | ||
NM_019636.3 | NP_062610.2 | 11 | 2105 | |||
GenBank mRNA | AK122445.1 | - | 11 | 2914 | ||
AK135665.1 | - | 10 | 1696 | |||
AK135892.1 | - | 10 | 1789 | |||
AK148356.1 | - | 10 | 1481 | |||
AK154591.1 | - | 12 | 2141 | |||
AK159256.1 | - | 11 | 2101 | |||
AK164923.1 | - | 7 | 809 | |||
BC059824.1 | - | 12 | 2470 | |||
BC117931.1 | - | 10 | 1773 | 40097712 | ||
U33005.1 | - | 11 | 2061 |