Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rqcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021383.5 | NP_067358.1 | 955 | |||
GenBank mRNA | AF221849.1 | - | 6 | 577 | ||
AK005025.2 | - | 6 | 957 | |||
BC050898.1 | - | 6 | 756 | 6401907 | ||
BC051948.1 | - | 794 | 5708192 |