Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021393.2 | NP_067368.2 | 6 | 1136 | ||
GenBank mRNA | AB041607.1 | - | 5 | 1257 | ||
AK080419.1 | - | 6 | 838 | |||
AK087524.1 | - | 6 | 836 | |||
AK173124.1 | - | 5 | 671 | |||
BC004762.1 | - | 6 | 1024 | 3155904 |