Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141933.1 | NP_001135405.1 | 2, 3 | 274 | ||
NM_021426.4 | NP_067401.3 | 2, 3 | 274 | |||
GenBank mRNA | AB030182.1 | - | 2, 3 | 252 | ||
AK005262.1 | - | 2, 3 | 245 | |||
AK007632.1 | - | 2, 3 | 239 | |||
AK021096.1 | - | 2, 3 | 269 | |||
BC061107.1 | - | 2, 3 | 259 | 30295742 | ||
BC089330.1 | - | 2, 3 | 226 | 30358178 | ||
CK625327.1 | - | 2, 3 | 274 | |||
EF058055.1 | - | 2, 3 | 199 | |||
EF058056.1 | - | 2, 3 | 199 | |||
EF058057.1 | - | 2, 3 | 199 |