Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021435.3 | NP_067410.1 | 7 | 750 | ||
GenBank mRNA | AB041549.1 | - | 7 | 751 | ||
AK029046.1 | - | 6 | 3371 | |||
AK029969.1 | - | 7 | 808 | |||
AK033913.1 | - | 7 | 680 | |||
AK042150.1 | - | 7 | 650 | |||
AK045791.1 | - | 7 | 693 | |||
AK138591.1 | - | 7 | 691 | |||
AK163299.1 | - | 2 | 218 | |||
BC016275.1 | - | 7 | 681 | |||
BC033512.1 | - | 7 | 675 | 4017336 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG062065.1 | - | 336 | Chr. - - [-] on Build |