Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021449.3 | NP_067424.2 | 11 | 1229 | ||
NM_175357.3 | NP_780566.1 | 11 | 1232 | |||
GenBank mRNA | AF229032.1 | - | 11 | 1184 | ||
AK032836.1 | - | 11 | 1206 | |||
AK050557.1 | - | 11 | 1215 | |||
AK075959.1 | - | 11 | 1205 | |||
AK076144.1 | - | 11 | 1212 | |||
AK077707.1 | - | 11 | 1212 | |||
AK160219.1 | - | 11 | 1208 | |||
BC033458.1 | - | 11 | 1190 | |||
BC046967.1 | - | 10 | 1147 | |||
BC069905.1 | - | 11 | 1189 | |||
BC086488.1 | - | 11 | 1207 | 30443656 |