Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021449.3 | NP_067424.2 | 5, 6 | 727 | ||
NM_175357.3 | NP_780566.1 | 5, 6 | 730 | |||
GenBank mRNA | AF229032.1 | - | 5, 6 | 682 | ||
AK032836.1 | - | 5, 6 | 704 | |||
AK050557.1 | - | 5, 6 | 713 | |||
AK075959.1 | - | 5, 6 | 704 | |||
AK076144.1 | - | 5, 6 | 710 | |||
AK077707.1 | - | 5, 6 | 710 | |||
AK160219.1 | - | 5, 6 | 706 | |||
BC033458.1 | - | 5, 6 | 688 | |||
BC046967.1 | - | 4, 5 | 645 | |||
BC069905.1 | - | 5, 6 | 687 | |||
BC086488.1 | - | 5, 6 | 705 | 30443656 |