Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153574.2 | NP_705802.1 | 11 | 1263 | ||
GenBank mRNA | AJ510136.1 | - | 11 | 1249 | ||
AK036291.1 | - | 4 | 470 | |||
AK052421.1 | - | 11 | 1204 | |||
BC005554.1 | - | 11 | 1168 | |||
BC054053.1 | - | 11 | 1237 | |||
BC060978.1 | - | 11 | 1200 | |||
BC080861.1 | - | 11 | 1263 | 30357292 |