Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021485.2 | NP_067460.1 | 7 | 560 | ||
GenBank mRNA | AJ007938.1 | - | 7 | 560 | ||
AK014412.1 | - | 7 | 639 | |||
AK090093.1 | - | 615 | ||||
AK156361.1 | - | 7 | 613 | |||
AK210365.1 | - | 3 | 212 | |||
BC051631.1 | - | 8 | 755 | |||
BC066842.1 | - | 7 | 631 | 30468251 |