Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021500.2 | NP_067475.2 | 3 | 302 | ||
GenBank mRNA | AF263247.1 | - | 3 | 264 | ||
AK007307.1 | - | 3 | 273 | |||
AK034859.1 | - | 3 | 301 | |||
AK049424.1 | - | 3 | 322 | |||
AK075637.1 | - | 2 | 142 | |||
AK088854.1 | - | 3 | 310 | |||
AK145800.1 | - | 3 | 365 | |||
AK152523.1 | - | 3 | 310 | |||
AK168788.1 | - | 3 | 328 | |||
AK169149.1 | - | 3 | 310 | |||
BC039054.1 | - | 3 | 288 | 2648667 | ||
BC058687.1 | - | 3 | 300 | 6411707 | ||
CX212584.1 | - | 3 | 302 |