Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngly1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021504.3 | NP_067479.2 | 9 | 1361 | ||
GenBank mRNA | AF250927.1 | - | 9 | 1348 | ||
AK028248.1 | - | 9 | 1362 | |||
AK148931.1 | - | 9 | 1364 | |||
BC028961.1 | - | 9 | 1315 | 5366023 |