Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021509.5 | NP_067484.2 | 3 | 650 | ||
GenBank mRNA | AB041606.1 | - | 3 | 585 | ||
AK014340.1 | - | 3 | 651 | |||
AK030274.1 | - | 3 | 591 | |||
AK081586.1 | - | 3 | 616 | |||
AK162030.1 | - | 3 | 592 |