Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021509.5 | NP_067484.2 | 5 | 858 | ||
GenBank mRNA | AB041606.1 | - | 5 | 793 | ||
AK014340.1 | - | 5 | 859 | |||
AK030274.1 | - | 5 | 799 | |||
AK081586.1 | - | 5 | 824 | |||
AK162030.1 | - | 5 | 800 | |||
BC025892.1 | - | 5 | 556 | |||
BC037684.1 | - | 5 | 796 | |||
BC057652.1 | - | 5 | 772 | 4017199 |