Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nampt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021524.2 | NP_067499.2 | 3 | 488 | ||
GenBank mRNA | AF234625.1 | - | 3 | 348 | ||
AK086415.1 | - | 3 | 319 | |||
AK153926.1 | - | 3 | 319 | |||
AK166270.1 | - | 3 | 348 | |||
AY679720.1 | - | 3 | 323 | |||
BC004059.1 | - | 3 | 296 | 3590087 | ||
BC018358.1 | - | 3 | 488 | 2609271 |