Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021526.2 | NP_067501.2 | 7 | 746 | ||
GenBank mRNA | AK011974.1 | - | 7 | 695 | ||
AK012013.1 | - | 7 | 677 | |||
AK150280.1 | - | 7 | 682 | |||
AK151104.1 | - | 7 | 729 | |||
AK151733.1 | - | 7 | 729 | |||
BC003742.1 | - | 7 | 651 | 3599671 | ||
Y13071.1 | - | 7 | 667 |