Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021531.6 | NP_067506.2 | 7, 8 | 1056 | ||
NM_153141.1 | NP_694781.1 | 7, 8 | 1056 | |||
GenBank mRNA | AF117887.2 | - | 7, 8 | 966 | ||
AK158242.1 | - | 7, 8 | 1021 | |||
AK158757.1 | - | 7, 8 | 896 | |||
BC003964.1 | - | 2, 3 | 163 | |||
BC036974.1 | - | 7, 8 | 1040 | 4935077 | ||
BC071267.1 | - | 8, 9 | 1172 |