Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163354.1 | NP_001156826.1 | 17 | 1694 | ||
NM_001163355.1 | NP_001156827.1 | 17 | 1694 | |||
NM_021536.7 | NP_067511.4 | 17 | 1694 | |||
GenBank mRNA | AF244542.1 | - | 4 | 328 | ||
AK019059.1 | - | 17 | 1654 | |||
AK028597.1 | - | 17 | 1637 | |||
AK031139.1 | - | 15 | 1356 | |||
AK032096.1 | - | 17 | 1667 | |||
AK041119.1 | - | 17 | 1630 | |||
AK054472.1 | - | 17 | 1665 | |||
AK078955.1 | - | 17 | 1637 | |||
AK090071.1 | - | 10 | 1020 | |||
AK138830.1 | - | 17 | 1635 | |||
AK141621.1 | - | 17 | 1521 | |||
AK147749.1 | - | 17 | 1673 | |||
AK147801.1 | - | 17 | 1699 | |||
AK167502.1 | - | 17 | 1694 | |||
BC006918.1 | - | 8 | 785 | |||
BC025021.1 | - | 3 | 807 | |||
BC044301.1 | - | 6 | 533 | |||
BC046785.1 | - | 17 | 1677 | 6416127 | ||
BC052882.1 | - | 7 | 638 | |||
BC058350.1 | - | 17 | 1609 | 6815832 |