Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021539.4 | NP_067514.2 | 4, 5 | 721 | ||
GenBank mRNA | AF033188.1 | - | 3, 4 | 565 | ||
AF072881.1 | - | 4, 5 | 642 | |||
AK136416.1 | - | 3, 4 | 621 | |||
AK150710.1 | - | 2, 3 | 295 | |||
BC055100.1 | - | 4, 5 | 721 | 6589250 |