Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Midn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB036882.1 | - | 4 | 816 | ||
AK154535.1 | - | 3 | 760 | |||
AK164324.1 | - | 4 | 834 | |||
AK164651.1 | - | 4 | 836 | |||
BC034719.1 | - | 4 | 786 | 3710931 | ||
BM944891.1 | - | 1 | 21 |