Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fign | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267846.1 | NP_001254775.1 | 4 | 2322 | ||
NM_001267847.1 | NP_001254776.1 | 4 | 2303 | |||
NM_021716.5 | NP_068362.1 | 3 | 2264 | |||
GenBank mRNA | AF263913.1 | - | 3 | 2214 | ||
AK143787.1 | - | 3 | 2239 | |||
AK163474.1 | - | 3 | 2235 | |||
AK164536.1 | - | 4 | 2275 | |||
BC059266.1 | - | 4 | 2322 | 6409940 |