Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021789.2 | NP_068561.1 | 3 | 639 | ||
GenBank mRNA | AF233340.1 | - | 3 | 596 | ||
AK005276.1 | - | 3 | 424 | |||
AK032168.1 | - | 3 | 405 | |||
BC038898.1 | - | 3 | 639 | 5401311 |