Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022019.5 | NP_071302.2 | 2 | 731 | ||
GenBank mRNA | AB037908.1 | - | 2 | 737 | ||
AK011995.1 | - | 2 | 715 | |||
AK035293.1 | - | 2 | 729 | |||
AK050528.1 | - | 2 | 729 | |||
AK088024.1 | - | 2 | 730 | |||
AK088186.1 | - | 2 | 732 | |||
AK088357.1 | - | 2 | 727 | |||
AK142568.1 | - | 2 | 733 | |||
BC025066.1 | - | 2 | 749 | 3495358 |