Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022319.2 | NP_071714.2 | 13 | 2258 | ||
GenBank mRNA | AJ278069.1 | - | 13 | 2206 | ||
AK048369.1 | - | 6 | 792 | |||
AK220500.1 | - | 13 | 2272 | |||
BC063058.1 | - | 13 | 2258 | 5705591 |