Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142939.1 | NP_001136411.1 | 2 | 544 | ||
GenBank mRNA | AK011927.1 | - | 2 | 529 | ||
AK019599.1 | - | 1407 | ||||
BC014715.1 | - | 2 | 493 | 4236212 |
Gene Symbol: Moap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142937.2 | NP_001136409.1 | 2 | 393 | ||
NM_022323.7 | NP_071718.1 | 2 | 437 | |||
GenBank mRNA | AU080670.1 | - | 2 | 377 | ||
BU614788.1 | - | 2 | 407 | |||
BY741099.1 | - | 2 | 391 |