Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 6 | 983 | ||
GenBank mRNA | AF136277.1 | - | 6 | 993 | ||
AF197479.1 | - | 6 | 910 | |||
AJ242663.1 | - | 6 | 993 | |||
AK002710.1 | - | 6 | 945 | |||
AK004095.1 | - | 6 | 981 | |||
AK008370.1 | - | 6 | 912 | |||
AK010912.1 | - | 6 | 979 | |||
AK150329.1 | - | 6 | 911 | |||
AK150894.1 | - | 6 | 918 | |||
AK150947.1 | - | 6 | 972 | |||
AK151440.1 | - | 6 | 922 | |||
AK151585.1 | - | 6 | 946 | |||
AK151994.1 | - | 6 | 956 | |||
AK153147.1 | - | 6 | 918 | |||
AK159118.1 | - | 6 | 929 | |||
AK159618.1 | - | 6 | 996 | |||
AK159826.1 | - | 6 | 916 | |||
AK160001.1 | - | 6 | 966 | |||
AK160028.1 | - | 6 | 966 | |||
AK160354.1 | - | 6 | 999 | |||
AK166047.1 | - | 6 | 967 | |||
AK170963.1 | - | 6 | 966 | |||
AK179924.1 | - | 2 | 209 | |||
AK185170.1 | - | 2 | 242 | |||
AK185852.1 | - | 2 | 243 | |||
AK192329.1 | - | 2 | 242 | |||
AK204750.1 | - | 2 | 184 | |||
AK205915.1 | - | 2 | 190 | |||
AK215212.1 | - | 2 | 242 | |||
BC008619.1 | - | 6 | 983 | 3588457 |