Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160180.1 | NP_001153652.1 | 3 | 1106 | ||
NM_001160181.1 | NP_001153653.1 | 3 | 1054 | |||
NM_001160182.1 | NP_001153654.1 | 3 | 985 | |||
NM_022329.4 | NP_071724.1 | 2 | 994 | |||
GenBank mRNA | AJ251363.1 | - | 2 | 961 | ||
AK003852.1 | - | 2 | 943 | |||
AK032697.1 | - | 2 | 938 | |||
AK075980.1 | - | 1 | 225 | |||
AK150117.1 | - | 3 | 1109 | |||
AK150303.1 | - | 3 | 1023 | |||
AK151453.1 | - | 3 | 1023 | |||
AK152137.1 | - | 3 | 1030 | |||
BC039927.1 | - | 2 | 959 | |||
BC053726.1 | - | 3 | 1009 | |||
BC060258.1 | - | 3 | 1335 | |||
BC064017.1 | - | 2 | 959 | 3995710 | ||
BC092059.1 | - | 3 | 1012 | 4019181 |