Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160180.1 | NP_001153652.1 | 3 | 951 | ||
NM_001160181.1 | NP_001153653.1 | 3 | 899 | |||
NM_001160182.1 | NP_001153654.1 | 3 | 830 | |||
NM_022329.4 | NP_071724.1 | 2 | 839 | |||
GenBank mRNA | AJ251363.1 | - | 2 | 806 | ||
AK003852.1 | - | 2 | 788 | |||
AK032697.1 | - | 2 | 783 | |||
AK075980.1 | - | 1 | 70 | |||
AK150117.1 | - | 3 | 954 | |||
AK150303.1 | - | 3 | 868 | |||
AK151453.1 | - | 3 | 868 | |||
AK152137.1 | - | 3 | 875 | |||
BC039927.1 | - | 2 | 804 | |||
BC053726.1 | - | 3 | 854 | |||
BC060258.1 | - | 3 | 1180 | |||
BC064017.1 | - | 2 | 804 | 3995710 | ||
BC092059.1 | - | 3 | 857 | 4019181 |