Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022331.1 | NP_071726.1 | 4 | 418 | ||
GenBank mRNA | AB034991.1 | - | 4 | 418 | ||
AK081688.1 | - | 4 | 422 | |||
AK159500.1 | - | 4 | 424 | |||
AK165832.1 | - | 4 | 424 | |||
AK172874.1 | - | 4 | 411 | |||
AK183178.1 | - | 2 | 78 | |||
BC013523.1 | - | 4 | 410 | 4236803 |