Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 3 | 499 | ||
NM_001252490.1 | NP_001239419.1 | 2 | 373 | |||
NM_001252492.1 | NP_001239421.1 | 2 | 742 | |||
NM_001252493.1 | NP_001239422.1 | 2 | 742 | |||
NM_207530.3 | NP_997413.2 | 16 | 1628 | |||
GenBank mRNA | AK050641.1 | - | 2 | 297 | ||
AK132676.1 | - | 3 | 498 | |||
AK161374.1 | - | 4 | 370 | |||
AK168366.1 | - | 2 | 703 | |||
AY536214.1 | - | 16 | 1507 | |||
BC015075.1 | - | 2 | 463 | |||
BC046466.1 | - | 2 | 505 | |||
BC076637.1 | - | 2 | 373 |