Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252489.1 | NP_001239418.1 | 5 | 733 | ||
NM_001252490.1 | NP_001239419.1 | 4 | 607 | |||
NM_001252491.1 | NP_001239420.1 | 3 | 813 | |||
NM_001252492.1 | NP_001239421.1 | 4 | 976 | |||
NM_001252493.1 | NP_001239422.1 | 4 | 976 | |||
NM_207530.3 | NP_997413.2 | 18 | 1862 | |||
GenBank mRNA | AB017026.1 | - | 2 | 857 | ||
AF394063.2 | - | 2 | 168 | |||
AK050641.1 | - | 4 | 531 | |||
AK132676.1 | - | 5 | 732 | |||
AK133806.1 | - | 2 | 239 | |||
AK161374.1 | - | 6 | 604 | |||
AK168366.1 | - | 4 | 937 | |||
AY536214.1 | - | 18 | 1741 | |||
BC015075.1 | - | 4 | 697 | |||
BC046466.1 | - | 4 | 739 | |||
BC057194.1 | - | 2 | 143 | |||
BC076637.1 | - | 4 | 607 | |||
BC092247.1 | - | 3 | 813 | |||
BY248497.1 | - | 2 | 451 | |||
BY249673.1 | - | 2 | 400 | |||
DV068702.1 | - | 2 | 203 |