Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146211.1 | NP_001139683.1 | 5 | 380 | ||
NM_001146212.1 | NP_001139684.1 | 5 | 383 | |||
NM_183086.2 | NP_898909.2 | 5 | 473 | |||
GenBank mRNA | AK004151.1 | - | 5 | 377 | ||
AK136970.1 | - | 1 | 505 | |||
BB862036.1 | - | 5 | 392 | |||
BC038917.1 | - | 5 | 369 | |||
BC048912.1 | - | 5 | 476 | 4238014 | ||
BC096043.1 | - | 4 | 338 | |||
BC096657.1 | - | 4 | 288 | 5144028 | ||
DV046794.1 | - | 5 | 394 |