Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022724.4 | NP_073561.2 | 4 | 1202 | ||
GenBank mRNA | AF149205.1 | - | 4 | 1180 | ||
AK015728.1 | - | 2 | 248 | |||
AK045206.1 | - | 3 | 1040 | |||
AK083457.1 | - | 4 | 1180 | |||
AK165278.1 | - | 3 | 855 | |||
BC032960.1 | - | 3 | 525 | 1330053 |
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK162838.1 | - | 1 | 1846 |