Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022814.2 | NP_073725.2 | 39 | 9665 | ||
GenBank mRNA | AF206329.1 | - | 39 | 9665 | ||
AK014693.1 | - | 2 | 549 | |||
AK136952.1 | - | 2 | 1936 | |||
BC008135.1 | - | 2 | 752 |