Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164885.1 | NP_001158357.1 | 3, 4 | 514 | ||
NM_022882.4 | NP_075020.2 | 3, 4 | 390 | |||
GenBank mRNA | AF286723.1 | - | 3, 4 | 390 | ||
AK033662.1 | - | 3, 4 | 427 | |||
AK050018.1 | - | 3, 4 | 515 | |||
AK086834.1 | - | 3, 4 | 326 | |||
AK136472.1 | - | 985 | ||||
BC039698.1 | - | 3, 4 | 503 | 5101211 |