Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022981.4 | NP_075357.4 | 8 | 1800 | ||
GenBank mRNA | AJ242914.1 | - | 8 | 1555 | ||
AK013583.1 | - | 8 | 1606 | |||
AK040989.1 | - | 8 | 1534 | |||
BC006657.1 | - | 8 | 1549 | 3500984 |