Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022981.4 | NP_075357.4 | 5 | 931 | ||
GenBank mRNA | AJ242914.1 | - | 5 | 686 | ||
AK013583.1 | - | 5 | 737 | |||
AK040989.1 | - | 5 | 665 | |||
BC006657.1 | - | 5 | 680 | 3500984 |