Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023044.2 | NP_075531.2 | 5 | 1718 | ||
GenBank mRNA | AF121080.1 | - | 5 | 1726 | ||
AK051941.1 | - | 5 | 1319 | |||
AK171698.1 | - | 1371 | ||||
BC051940.1 | - | 5 | 1389 | 5717926 |