Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023906.3 | NP_076395.2 | 8 | 1216 | ||
GenBank mRNA | AF155354.1 | - | 7 | 1031 | ||
AK010315.1 | - | 8 | 1152 | |||
AK028489.1 | - | 9 | 1285 | |||
AK038086.1 | - | 8 | 1197 | |||
AK143580.1 | - | 8 | 1151 | |||
BC023086.1 | - | 8 | 1150 | 4947610 |