Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clstn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290989.1 | NP_001277918.1 | 9 | 1652 | ||
NM_023051.5 | NP_075538.1 | 10 | 1682 | |||
GenBank mRNA | AJ289016.1 | - | 10 | 1513 | ||
AK034544.1 | - | 2 | 225 | |||
AK173059.1 | - | 9 | 1692 | |||
BC029027.1 | - | 5 | 767 | |||
BC053843.1 | - | 9 | 1621 | 6315355 | ||
CO433558.1 | - | 4 | 655 |