Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023053.3 | NP_075540.1 | 5 | 773 | ||
GenBank mRNA | AF292033.1 | - | 712 | |||
AJ297390.1 | - | 5 | 723 | |||
AK018524.1 | - | 1 | 140 | |||
AK033504.1 | - | 5 | 732 | |||
AK049646.1 | - | 5 | 767 | |||
AK168547.1 | - | 5 | 723 | |||
AK168633.1 | - | 5 | 732 | |||
AK191126.1 | - | 1 | 86 | |||
AK198217.1 | - | 1 | 65 | |||
AK200138.1 | - | 1 | 77 | |||
BC004850.1 | - | 5 | 728 | 2810960 |