Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 5 | 1112 | ||
GenBank mRNA | AF268871.1 | - | 5 | 1076 | ||
AF283518.1 | - | 5 | 1067 | |||
AK028424.1 | - | 5 | 1094 | |||
AK087501.1 | - | 5 | 1100 | |||
AK147924.1 | - | 5 | 1081 | |||
AK171885.1 | - | 5 | 1092 | |||
AK184409.1 | - | 2 | 334 | |||
AK186069.1 | - | 1 | 307 | |||
BC024915.1 | - | 5 | 1086 | 3158026 | ||
BC099675.1 | - | 5 | 1070 | 5011896 |