Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eefsec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023060.3 | NP_075547.1 | 2 | 529 | ||
GenBank mRNA | AF268871.1 | - | 2 | 493 | ||
AF283518.1 | - | 2 | 484 | |||
AK028424.1 | - | 2 | 511 | |||
AK087501.1 | - | 2 | 517 | |||
AK147924.1 | - | 2 | 498 | |||
AK171885.1 | - | 2 | 509 | |||
AK207457.1 | - | 1 | 197 | |||
AK216395.1 | - | 1 | 131 | |||
BC024915.1 | - | 2 | 503 | 3158026 | ||
BC099675.1 | - | 2 | 487 | 5011896 |