Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025314.3 | NP_079590.1 | 3 | 401 | ||
GenBank mRNA | AK002246.1 | - | 3 | 337 | ||
AK003594.1 | - | 3 | 373 | |||
AK010822.1 | - | 3 | 339 | |||
AK011917.1 | - | 3 | 369 | |||
AK158898.1 | - | 3 | 458 | |||
BC026537.1 | - | 3 | 374 | 5008303 |