Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025314.3 | NP_079590.1 | 3 | 447 | ||
GenBank mRNA | AK002246.1 | - | 3 | 383 | ||
AK003594.1 | - | 3 | 419 | |||
AK010822.1 | - | 3 | 385 | |||
AK011917.1 | - | 3 | 415 | |||
AK017565.1 | - | 3 | 371 | |||
AK158898.1 | - | 3 | 504 | |||
BC026537.1 | - | 3 | 420 | 5008303 |