Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252444.1 | NP_001239373.1 | 7, 8 | 467 | ||
NM_001252445.1 | NP_001239374.1 | 7, 8 | 467 | |||
NM_144954.3 | NP_659203.1 | 7, 8 | 467 | |||
GenBank mRNA | AK009460.1 | - | 7, 8 | 428 | ||
AK012472.1 | - | 7, 8 | 448 | |||
AK081792.1 | - | 7, 8 | 449 | |||
AK090052.1 | - | 7, 8 | 460 | |||
AK161596.1 | - | 7, 8 | 452 | |||
BC028899.1 | - | 7, 8 | 428 | 4021590 |