Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 10 | 1438 | ||
NM_023149.3 | NP_075638.2 | 11 | 1406 | |||
GenBank mRNA | AB046738.1 | - | 11 | 1347 | ||
AK003779.1 | - | 11 | 1327 | |||
AK131124.1 | - | 10 | 1317 | |||
AK149751.1 | - | 10 | 1362 | |||
AK151013.1 | - | 11 | 1328 | |||
AK152442.1 | - | 10 | 1403 | |||
AK152729.1 | - | 10 | 1376 | |||
AK153040.1 | - | 10 | 1377 | |||
AK159129.1 | - | 10 | 1405 | |||
AK159739.1 | - | 11 | 1333 | |||
AK168029.1 | - | 11 | 1345 | |||
AK168652.1 | - | 11 | 1357 | |||
AK168856.1 | - | 9 | 1095 | |||
AK172034.1 | - | 11 | 1328 | |||
AK185765.1 | - | 2 | 90 | |||
AK208750.1 | - | 1 | 41 | |||
AK214219.1 | - | 2 | 90 | |||
BC005532.1 | - | 11 | 1337 | 3496319 |