Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fopnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025345.2 | NP_079621.1 | 2 | 146 | ||
GenBank mRNA | AK012206.1 | - | 2 | 141 | ||
AK146884.1 | - | 2 | 172 | |||
AK153894.1 | - | 2 | 150 | |||
BC011300.1 | - | 2 | 146 | 3671642 |