Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025386.3 | NP_079662.1 | 2 | 156 | ||
GenBank mRNA | AK002325.1 | - | 2 | 136 | ||
AK003856.1 | - | 2 | 156 | |||
AK008324.1 | - | 2 | 153 | |||
AK010335.1 | - | 2 | 969 | |||
BC049581.1 | - | 2 | 133 | 6772363 | ||
BY708252.1 | - | 2 | 153 |