Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025399.3 | NP_079675.1 | 5 | 500 | ||
GenBank mRNA | AK003991.1 | - | 5 | 507 | ||
AK013174.1 | - | 5 | 456 | |||
BC025444.1 | - | 5 | 502 | 3986887 |