Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025418.3 | NP_079694.2 | 5 | 475 | ||
GenBank mRNA | AK002611.1 | - | 5 | 474 | ||
AK003213.1 | - | 4 | 392 | |||
AK004323.1 | - | 5 | 547 | |||
AK009227.2 | - | 5 | 464 | |||
BC026752.1 | - | 5 | 468 | 4034998 | ||
CX212626.1 | - | 5 | 476 |